missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin8 Antibody (mAbGPa) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53121-0.025mg
This item is not returnable.
View return policy
Description
Actin8 Monoclonal specifically detects Actin8 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.
Specifications
| Actin8 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence | |
| ACT8, Actin-8 | |
| Purified Arabidopsis Actin8 | |
| 0.025 mg | |
| Apoptosis, Cancer, Cellular Signaling | |
| This mouse monoclonal antibody was generated against plant actin ACT8 and recognizes all tested subclasses of plant actins (arabidopsis ACT1, 2, 3, 4, 7, 8, 11, 12) and Dictyosetelium actin. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| mAbGPa | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Plant | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction