missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Actinomycin D, 93%

Product Code. 10043673 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
Packaging:
Glass Bottle
Unit Size:
100mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10043673 1 mg Glass Bottle 1mg
10247270 5 mg Glass Bottle 5mg
10308880 25 mg Glass Bottle 25mg
10413075 100 mg Glass Bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10043673 Supplier Thermo Scientific Chemicals Supplier No. 294940010
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 50-76-0 | C62H86N12O16 | 1255.438 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.438
MDL Number MFCD00005033
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
Synonym Dactinomycin
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C

Specifications

Type Actinomycin D
Physical Form Crystalline Powder
CAS Min % 91.0
CAS Max % 100.0
Melting Point 245.0°C to 252.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Color Red
Quantity 1 mg
Assay Percent Range 93%
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, 1,2-propanediol, water + glyco, l mixture
Formula Weight 1255.44
Percent Purity 93%
Chemical Name or Material Actinomycin D, 93%
Show More Show Less
compliance-icons
Product Identifier
  • Actinomycin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.