missing translation for 'onlineSavingsMsg'
Learn More

Actinomycin D, >99%, MP Biomedicals™

Product Code. 11477571 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477571 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477571 Supplier MP Biomedicals Supplier No. 0210465801
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-76-0 | Formula: C62H86N12O16 | Weight: 1255.438 g/mol

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.438
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
Synonym Dactinomycin, Actinomycin-[thr-val-pro-sar-meval]
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C

Specifications

Type Actinomycin D
Physical Form Powder
Quantity 1 mg
Assay Percent Range >99%
Formula Weight 1255.5
Grade Not Available
Chemical Name or Material Actinomycin D
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.