missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Actinomycin D ReadyMade™ Solution

Product Code. 30044027
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044027 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044027 Supplier TOKU-E™ Supplier No. A22710x1ML
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Actinomycin D ReadyMade™ Solution is provided as a filter-sterilized solution of Actinomycin D in DMSO.

  • CAS Number: 50-76-0
  • Molecular Formula: C62H86N12O16
  • Molecular Weight: 1255.42 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomyces sp.
  • Spectrum: Actinomycin D has an inhibitory effect on Gram-positive, Gram-negative bacteria and some fungi. However, its toxicity precludes its use for infectious diseases
  • Mechanism of Action: Actinomycin is a cyclic polypeptide-containing antibiotic that binds to DNA and inhibits RNA polymerase activity

As Actinomycin can bind DNA duplexes, it can also interfere with DNA replication, although other chemicals such as hydroxyurea are better suited for use in the laboratory as inhibitors of DNA synthesis. Actinomycin D and its fluorescent derivative,7-aminoactinomycin D (7-AAD), are used as stains in microscopy and flow cytometry applications. The affinity of these stains/compounds for GC-rich regions of DNA strands makes them excellent markers for DNA. 7-AAD binds to single stranded DNA; therefore, it is a useful tool in determining apoptosis and distinguishing between living and dead cells.

Specifications

CAS 50-76-0
Certifications/Compliance Purity >95.0%
Chemical Name or Material Actinomycin D
Color Clear to Dark orange
Content And Storage -20°C, Protect from light
Description Actinomycin ReadyMade™ Solution 2 mg/mL in DMSO, Sterile Filtered
Concentration 2.0 to 2.1 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid, 0.2% in Dimethyl Sulfoxide
For Use With (Application) Microbiology, Plant Biology, Eucaryotic Cell Culture and Cancer Applications
Manufacturing Quality 1 mL
Product Line ReadyMade™ Solutions
Purity or Quality Grade >95.0%
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Source Streptomyces sp.
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.