missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR3 Antibody (3G2C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00 € - 562.00 €
Specifications
| Antigen | ACTR3 |
|---|---|
| Clone | 3G2C9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18223244
|
Novus Biologicals
NBP2-61806 |
0.1 mL |
562.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668786
|
Novus Biologicals
NBP2-61806-0.025ml |
0.025 mL |
203.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACTR3 Monoclonal specifically detects ACTR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACTR3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| actin-related protein 3, ARP3 (actin-related protein 3, yeast) homolog, ARP3 actin-related protein 3 homolog (yeast), ARP3Actin-like protein 3 | |
| ACTR3 | |
| IgG1 | |
| Protein G purified |
| 3G2C9 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Human, Mouse | |
| 10096 | |
| Purified recombinant fragment of human ACTR3 (AA: 287-418) expressed in E. Coli. | |
| Primary | |
| 47.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title