missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acylglycerol Kinase Antibody (9A3B1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05339-100ul
This item is not returnable.
View return policy
Description
Acylglycerol Kinase Monoclonal antibody specifically detects Acylglycerol Kinase in Human samples. It is validated for Western Blot
Specifications
| Acylglycerol Kinase | |
| Monoclonal | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 9A3B1 | |
| Western Blot 1:500-1:2000 | |
| acylglycerol kinase, EC 2.7.1.107, EC 2.7.1.94, FLJ10842, hAGK, HsMuLK, mitochondrial, MuLK, MULKhsMuLK, Multi-substrate lipid kinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-422 of human Acylglycerol Kinase (NP_060708.1). MTVFFKTLRNHWKKTTAGLCLLTWGGHWLYGKHCDNLLRRAACQEAQVFGNQLIPPNAQVKKATVFLNPAACKGKARTLFEKNAAPILHLSGMDVTIVKTDYEGQAKKLLELMENTDVIIVAGGDGTLQEVVTGVLRRTDEATFSKIPIGFIPLGETSSLSHTLFAESGNKVQHITDATLAIVKGETVPLDVLQIKGEKEQPVFAMTGLRWGSFRDAGVKVSKYWYLGPLKIKAAHFFSTLKEWPQTHQASISYTGPTERPPNEPEETPVQRPSLYRRILRRLASYWAQPQDALSQEVSPEVWKDVQLSTIELSITTRNNQLDPTSKEDFLNICIEPDTISKGDFITIGSRKVRNPKLHVEGTECLQASQCTLLIPEGAGGSFSIDSEEYEAMPVEVKLLPRKLQFFCDPRKREQMLTSPTQ | |
| 100 μg | |
| Signal Transduction | |
| 55750 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction