missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acylglycerol Kinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49085
This item is not returnable.
View return policy
Description
Acylglycerol Kinase Polyclonal antibody specifically detects Acylglycerol Kinase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Acylglycerol Kinase | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| acylglycerol kinase, EC 2.7.1.107, EC 2.7.1.94, FLJ10842, hAGK, HsMuLK, mitochondrial, MuLK, MULKhsMuLK, Multi-substrate lipid kinase | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IVKGETVPLDVLQIKGEKEQPVFAMTGLRWGSFRDAGVKVSKYWYLGPLKIKAAHFFSTLKEWPQTHQASISYTGPTERPPNEPEETPVQRPSLYRRILRRLASYWAQ | |
| 0.1 mL | |
| Signal Transduction | |
| 55750 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction