missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAM22 Recombinant Mouse Monoclonal Antibody (K40080_20C9)
GREENER_CHOICE

Product Code. 30827815
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827815 100 μL
30830371 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827815 Supplier Invitrogen™ Supplier No. MA564466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: QGDYVKKPGD GDSFYSDIPP GVSTNSASSS KKRSNGLSHS WSERIPDTKH ISDICENGRP RSNSWQGNLG GNKKKIRGKR FRPRSNSTET LSPAKSPSSS TGSIASSRKY PYPMPPLPDE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Pig - 99%, Cynomolgus monkey - 100%.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is highly expressed in the brain and may function as an integrin ligand in the brain. Alternative splicing results in several transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM22
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K40080_20C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene Adam22
Gene Accession No. Q9P0K1, Q9R1V6
Gene Alias 2900022I03Rik; a disintegrin and metallopeptidase domain 22; a disintegrin and metalloprotease domain (ADAM) 22; a disintegrin and metalloprotease domain 22; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 22; ADAM; ADAM 22; ADAM metallopeptidase domain 22; Adam22; ADAMs; AI854032; Disintegrin and metalloproteinase domain-containing protein 22; MDC2; metalloendopeptidases; Metalloproteinase-disintegrin ADAM22-3; metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2; MGC149832
Gene Symbols Adam22
Host Species Mouse
Immunogen Recombinant human ADAM22 (aa 771-890).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11496, 53616
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.