missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM28 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | ADAM28 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18221963
|
Novus Biologicals
NBP2-55995 |
100 μL |
624.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605687
|
Novus Biologicals
NBP2-55995-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAM28 Polyclonal specifically detects ADAM28 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ADAM28 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 10863 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ELPGVKKYEVVYPIRLHPLHKREAKEPEQQEQFETELKYKMTINGKIAVLYLKKNKNLLAPGYTETYYNSTGKEITTSPQIMDDCYY | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L | |
| ADAM28 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title