missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMDEC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92234-0.1ml
This item is not returnable.
View return policy
Description
ADAMDEC1 Polyclonal antibody specifically detects ADAMDEC1 in Human samples. It is validated for Western Blot
Specifications
| ADAMDEC1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| A disintegrin and metalloproteinase domain-like protein decysin-1, ADAM DEC1, ADAM-like protein decysin-1, ADAM-like, decysin 1, decysin, disintegrin protease, EC 3.4.24.-, FLJ79219, M12.219 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 206-470 of human ADAMDEC1 (NP_055294.1). KSPEKEDFLRAQKYIDLYLVLDNAFYKNYNENLTLIRSFVFDVMNLLNVIYNTIDVQVALVGMEIWSDGDKIKVVPSASTTFDNFLRWHSSNLGKKIHDHAQLLSGISFNNRRVGLAASNSLCSPSSVAVIEAKKKNNVALVGVMSHELGHVLGMPDVPFNTKCPSGSCVMNQYLSSKFPKDFSTSCRAHFERYLLSQKPKCLLQAPIPTNIMTTPVCGNHLLEVGEDCDCGSPKECTNLCCEALTCKLKPGTDCGGDAPNHTTE | |
| 0.1 mL | |
| Signal Transduction | |
| 27299 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction