missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTSL-1/Punctin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92531-0.1ml
This item is not returnable.
View return policy
Description
ADAMTSL-1/Punctin Polyclonal antibody specifically detects ADAMTSL-1/Punctin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ADAMTSL-1/Punctin | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ADAM-TS related protein 1, ADAMTSL-1, ADAMTS-like 1, ADAMTS-like protein 1, ADAMTSR1MGC118805, C9orf94, chromosome 9 open reading frame 94, DKFZp686L03130, FLJ35283, FLJ41032, FLJ46891, MGC118803, MGC40193, PUNCTIN, Punctin-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1483-1762 of human ADAMTSL1 (NP_001035362.3). SLVIQDYWWSVDRLATCSASCGNRGVQQPRLRCLLNSTEVNPAHCAGKVRPAVQPIACNRRDCPSRWMVTSWSACTRSCGGGVQTRRVTCQKLKASGISTPVSNDMCTQVAKRPVDTQACNQQLCVEWAFSSWGQCNGPCIGPHLAVQHRQVFCQTRDGITLPSEQCSALPRPVSTQNCWSEACSVHWRVSLWTLCTATCGNYGFQSRRVECVHARTNKAVPEHLCSWGPRPANWQRCNITPCENMECRDTTRYCEKVKQLKLCQLSQFKSRCCGTCGKA | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 92949 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction