missing translation for 'onlineSavingsMsg'
Learn More

Adapalene, MedChemExpress

Product Code. 16442006
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442006 500 mg 500mg
16434227 50 mg 50mg
16428086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16442006 Supplier Medchem Express Supplier No. HYB0091
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Adapalene (CD271), a third-generation synthetic retinoid, is widely used for the research of acne. Adapalene is a potent RAR agonist, with AC50s of 2.3 nM, 9.3 nM, and 22 nM for RARβ, RARγ, RARα, respectively. Adapalene also inhibits the enzymatic activity of GOT1 in a non-competitive manner. Adapalene exhibits anti-tumor activity.

Adapalene (CD271), a third-generation synthetic retinoid, is widely used for the research of acne. Adapalene is a potent RAR agonist, with AC50s of 2.3 nM, 9.3 nM, and 22 nM for RARβ, RARγ, RARα, respectively. Adapalene also inhibits the enzymatic activity of GOT1 in a non-competitive manner. Adapalene exhibits anti-tumor activity.

Specifications

Chemical Name or Material Adapalene
CAS 106685-40-9
Color White
Purity Grade Notes Research
Molecular Formula C28H28O3
Quantity 500 mg
Synonym CD271
Solubility Information DMSO : 20 mg/mL (48.48 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=C(C23CC4CC(C3)CC(C2)C4)C=C(C5=CC6=C(C=C(C(O)=O)C=C6)C=C5)C=C1
Molecular Weight (g/mol) 412.52
Formula Weight 412.52
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.