Learn More
Memmert™ Additional Stainless Steel Grid for SF260plus, Reinforced Chamber Option
Product Code. 17548205 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF260plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17548205 SF260plus Sterilizer Each
1 options
Product Code. 17548205 Supplier Memmert™ Supplier No. SF260plusE29766
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Additional grid features a sturdy stainless steel construction.
  • Reinforced
  • Stainless Steel
  • Electropoloished
  • Max. loading 60 kg
Specifications
For Use With (Equipment) SF260plus Sterilizer
Product Type Reinforced Grid
Material Stainless Steel