missing translation for 'onlineSavingsMsg'
Learn More

Adenine, MedChemExpress

Product Code. 16412069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412069 1 g 1g
16400798 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16412069 Supplier Medchem Express Supplier No. HYB0152
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Adenine (6-Aminopurine), a purine, is one of the four nucleobases in the nucleic acid of DNA. Adenine acts as a chemical component of DNA and RNA. Adenine also plays an important role in biochemistry involved in cellular respiration, the form of both ATP and the cofactors (NAD and FAD), and protein synthesis.

Adenine (6-Aminopurine), a purine, is one of the four nucleobases in the nucleic acid of DNA.

Specifications

Chemical Name or Material Adenine
Physical Form Solid
CAS 73-24-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C5H5N5
For Use With (Application) Cancer-programmed cell death
Synonym 6-Aminopurine Vitamin B4
Solubility Information DMSO : 20 mg/mL (148.01 mM; Need ultrasonic) ∣1M HCl : 20 mg/mL (148.01 mM; Need ultrasonic) ∣H2O : 2 mg/mL (14.80 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=C2N=CNC2=NC=N1
Molecular Weight (g/mol) 135.13
Formula Weight 135.13
Percent Purity 99.14%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.