missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine Deaminase/ADA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98772-100ul
This item is not returnable.
View return policy
Description
Adenosine Deaminase/ADA Polyclonal antibody specifically detects Adenosine Deaminase/ADA in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Adenosine Deaminase/ADA | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:500-1:3000 | |
| ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Adenosine Deaminase/ADA. | |
| 100 μg | |
| Cancer, Endocrinology | |
| 100 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction