missing translation for 'onlineSavingsMsg'
Learn More

Adenosine, TRC

Product Code. 30011412
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011412 1 g 1g
30010871 50 mg 50mg
30007967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011412 Supplier Toronto Research Chemicals Supplier No. TRCA2804001G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10 H13 N5 O4
Molecular Weight (g/mol) 267.24
Synonym 9-β-D-Ribofuranosyl-9H-purin-6-amine, 9-β-D-Ribofuranosyladenine, 9H-Purin-6-amine, 9-β-D-ribofuranosyl-, Adenine riboside, Adenocard, Adenocor, Adenogesic, Adenoscan, Adrekar, Boniton, D-Adenosine, Myocol, NSC 7652, Nucleocardyl, Riboadenosine, Sandesin, β-Adenosine, β-D-Adenosine, β-D-Ribofuranose, 1-(6-amino-9H-purin-9-yl)-1-deoxy-, β-D-Ribofuranoside, adenine-9
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES Nc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

Specifications

Analyte or Component Names Adenosine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 1 g
Formula Weight 267.0968
Percent Purity >95%
Chemical Name or Material Adenosine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.