missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Adenovirus Expression Medium (AEM) is recommended for the growth of PER.C6™ cells in suspension and for the production of viruses and viral vectors in these cells. It is an animal-origin free cell culture medium without components directly derived from human or animal sources. Each lot of AEM is performance tested for cell growth with PER.C6™ cells and AEM has been qualified to produce adenovirus and influenza viruses. It also supports HEK 293 cell growth. Glutamine or GlutaMAX™-I supplementation is required.
Specifications
Specifications
| Cell Line | 293 (HEK), 293 (E1), PER.C6 |
| Classification | Animal Origin-free, Serum-free |
| Form | Liquid |
| Product Type | N-Terminal Fusion Kit |
| Serum Level | Serum-free |
| Without Additives | No Glutamine |
| Product Line | Gibco |
| Quantity | 1000 mL |
| Shipping Condition | Room Temperature |
| Content And Storage | Store at 2°C to 8°C, in the dark. |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals..
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?