missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-92683
This item is not returnable.
View return policy
Description
Adenylate Cyclase 3 Polyclonal specifically detects Adenylate Cyclase 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Adenylate Cyclase 3 | |
| Polyclonal | |
| Unconjugated | |
| 50% PBS, 50% Glycerol with 5mM Sodium Azide | |
| AC3EC 4.6.1.1, AC-III, adenylate cyclase 3, adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, adenylyl cyclase, type III, ATP pyrophosphate-lyase 3, EC 4.6.1, KIAA0511, olfactive type | |
| Rabbit | |
| 180 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience | |
| 109 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:10000-1:20000, Immunocytochemistry/Immunofluorescence 1:10000-1:20000 | |
| O60266 | |
| ADCY3 | |
| C-terminal peptide of rat ACIII, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction