missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 3 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05526-100ul
This item is not returnable.
View return policy
Description
Adenylate Cyclase 3 Monoclonal antibody specifically detects Adenylate Cyclase 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Adenylate Cyclase 3 | |
| Monoclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| AC3EC 4.6.1.1, AC-III, adenylate cyclase 3, adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, adenylyl cyclase, type III, ATP pyrophosphate-lyase 3, EC 4.6.1, KIAA0511, olfactive type | |
| C-terminal peptide of rat Adenylate Cyclase 3, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH. | |
| 100 μg | |
| Lipid and Metabolism, Neuroscience | |
| 109 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction