missing translation for 'onlineSavingsMsg'
Learn More

Adezmapimod hydrochloride, MedChemExpress

Product Code. 16484806
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484806 100 mg 100mg
16450127 50 mg 50mg
16423748 10 mg 10mg
16410966 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484806 Supplier Medchem Express Supplier No. HY10256A
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Adezmapimod (SB 203580) hydrochloride is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. Adezmapimod hydrochloride inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. Adezmapimod hydrochloride is an autophagy and mitophagy activator.

Adezmapimod (SB 203580) hydrochloride is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. Adezmapimod hydrochloride inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. Adezmapimod hydrochloride is an autophagy and mitophagy activator.

Specifications

Chemical Name or Material Adezmapimod hydrochloride
CAS 869185-85-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H17ClFN3OS
Quantity 100 mg
Synonym SB 203580 hydrochlorideRWJ 64809 hydrochloride
Solubility Information DMSO : 100 mg/mL (241.60 mM; Need ultrasonic) ∣H2O : 8.43 mg/mL (20.37 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(C2=NC(C3=CC=C(F)C=C3)=C(C4=CC=NC=C4)N2)C=C1)C.[H]Cl
Molecular Weight (g/mol) 413.9
Formula Weight 413.9
Percent Purity 98.6%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.