missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Adflo™ Gas Filter, A1B1E1, 837242
Click to view available options
:
Category: Welding Parts and Accessories, Model: 83 72 42
Description
- Provides protection against a range of particulate and gas hazards (dependent upon filters selected)
- For use with 3M™ Adflo™ Powered Air Purifying Respirators (PAPR)
- Particle filter, gas filters, and odour filter/pad available
- Installation is easy and secure
- For highest possible safety and comfort, regularly check your equipment for worn or damaged parts
Specifications
Specifications
| Cartridge Type | Organic Vapor, Acid Gas |
| Color | Black |
| Product Type | Filter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction