missing translation for 'onlineSavingsMsg'
Learn More

Color Coded High Resolution pH Buffer, Reagecon™
BIOPHARMA

Product Code. 11778465 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
pH:
4
4.00
7
7.00
10.00
Unit Size:
500mL
Each
This item is not returnable. View return policy

Product Code. 11778465

Brand: Reagecon 110000C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use color coded pH buffers for ease of use. Reagecon™ Color Coded High Resolution pH Buffers are tested at either 20°C or 25°C and are certified and NIST traceable.

  • Colored pH Buffer Solutions
  • High Resolution
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes

Specifications

Melting Point 0°C
Color Blue
pH 10.00
Physical Form Liquid
Quantity 500 mL
Odor Characteristic
Solubility Information Fully miscible in water
compliance-icons
Product Identifier
  • BUFFER SOLUTION pH 10.000 ± 0.010 @ 20°C
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Boric Acid, DisodiumSalt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.