missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin Mouse, Clone: ADPN/1370, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 9370-MSM3-P0
This item is not returnable.
View return policy
Specifications
| Adiponectin | |
| Cytoplasmic and Cell Surface | |
| ADPN/1370 | |
| Unconjugated | |
| NP_001171271 | |
| Mouse | |
| 26-30kDa | |
| Protein A/G Purified | |
| RUO | |
| 9370 | |
| IgG2b |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| ADIPOQ | |
| Recombinant human Adiponectin protein fragment | |
| HeLa cells or Kidney | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction