missing translation for 'onlineSavingsMsg'
Learn More

Adjudin, MedChemExpress

Product Code. 16497678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497678 10 mg 10mg
16477576 100 mg 100mg
16463687 50 mg 50mg
16410148 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497678 Supplier Medchem Express Supplier No. HY18996
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Adjudin is an extensively studied male contraceptive with a superior mitochondria-inhibitory effect. Adjudin is also a potent Cl- channel blocker.

Adjudin is an extensively studied male contraceptive with a superior mitochondria-inhibitory effect. Adjudin is also a potent Cl- channel blocker.

Specifications

Chemical Name or Material Adjudin
Physical Form Solid
CAS 252025-52-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H12Cl2N4O
For Use With (Application) Cancer-programmed cell death
Synonym AF-2364
Solubility Information DMSO : 16.67 mg/mL (49.73 mM; Need ultrasonic)
SMILES O=C(C1=NN(CC2=CC=C(Cl)C=C2Cl)C3=C1C=CC=C3)NN
Molecular Weight (g/mol) 335.19
Formula Weight 335.19
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.