missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | ADK |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617655
|
Novus Biologicals
NBP3-00077-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601606
|
Novus Biologicals
NBP3-00077-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADK Polyclonal antibody specifically detects ADK in Human samples. It is validated for Western Blot, ELISASpecifications
| ADK | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 132 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
| Produced in rabbits immunized with purified, recombinant Human ADK (Accession#: AAH03568.1; Met 1-His 345). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title