missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adrenomedullin/ADM Antibody (HTA 91a/G2), Novus Biologicals™
Mouse Monoclonal Antibody
188.00 € - 463.00 €
Specifications
| Antigen | Adrenomedullin/ADM |
|---|---|
| Clone | HTA 91a/G2 |
| Concentration | 2 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18586961
|
Novus Biologicals
NB100-2632-0.025ml |
0.025 mL |
188.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18551062
|
Novus Biologicals
NB100-2632-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adrenomedullin/ADM Monoclonal antibody specifically detects Adrenomedullin/ADM in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Adrenomedullin/ADM | |
| 2 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience | |
| PBS | |
| 133 | |
| IgM | |
| Thiophilic Chromatography |
| HTA 91a/G2 | |
| Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| adrenomedullin, AMpreproadrenomedullin | |
| Chemically synthesized full length adrenomedullin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title