missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adrenomedullin R/ADMR/GPR182 Antibody (S02-3E6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19650-100ul
This item is not returnable.
View return policy
Description
Adrenomedullin R/ADMR/GPR182 Monoclonal antibody specifically detects Adrenomedullin R/ADMR/GPR182 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Adrenomedullin R/ADMR/GPR182 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| S02-3E6 | |
| Unconjugated | |
| ADMR, adrenomedullin receptor, AMR, AM-R, G protein-coupled receptor 182,7TMR, G10D, gamrh, G-protein coupled receptor 182, hrhAMR, MGC34399 | |
| Recombinant protein of human Adrenomedullin R/ADMR/GPR182 (Uniprot # O15218) | |
| 100 μg | |
| GPCR | |
| 11318 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction