missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adrenomedullin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10778-1-AP-20UL
This item is not returnable.
View return policy
Description
Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintece of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.Specifications
| Adrenomedullin | |
| Polyclonal | |
| Unconjugated | |
| ADM | |
| ADM, adrenomedullin, AM, PAMP, ProAM N terminal 20 peptide, ProAM N20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 133 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35318 | |
| ADM | |
| Adrenomedullin Fusion Protein Ag1197 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction