missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADSL Monoclonal Antibody (OTI2D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501810
This item is not returnable.
View return policy
Description
Adenylosuccinate lyase catalyzes two non-sequential steps in de novo AMP synthesis: converts (S)-2-(5-amino-1-(5-phospho-D-ribosyl)imidazole-4-carboxamido)succinate (SAICAR) to fumarate plus 5-amino-1-(5-phospho-D-ribosyl)imidazole-4-carboxamide, and thereby also contributes to de novo IMP synthesis, and converts succinyladenosine monophosphate (SAMP) to AMP and fumarate.Specifications
| ADSL | |
| Monoclonal | |
| 0.55 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30566 | |
| ADSL | |
| Full length human recombit protein of human ADSL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D10 | |
| Unconjugated | |
| ADSL | |
| AMPS, ASASE, ASL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 158, 315150, 474499 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction