missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Adult Normal Kidney Slides (Adult Normal)- Paraffin

Product Code. 18724933 Shop All Bio Techne Products
Change view
Click to view available options
Species:
Human
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species Unit Size
18724933 Human Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18724933 Supplier Novus Biologicals™ Supplier No. NBP178269
  • ONLINE EXCLUSIVE PRICE
    447.00 €
    422.10 € / Pack of 5
    Save 24.90 €
    5% Off
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type Adult Normal Kidney Slides (Adult Normal)
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.