missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AEC Chromogen/Substrate

Product Code. 18784953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18784953 30 mL 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18784953 Supplier R&D Systems™ Supplier No. NB90079773
  • ONLINE EXCLUSIVE PRICE
    483.93 €
    392.70 € / 30mL
    Save 91.23 €
    18% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Novus
Quantity 30 mL
For Use With (Application) Stable Chromogen/Substrate Solution to be Used in Conjunction with Peroxidase-Based Immunostaining Systems. Specimens Stained Using AEC Chromogen/Substrate Cannot be Dehydrated in Ethanol and Hence Need to be Mounted in an Aqueous Based Mounting Medium. Staining Procedure 1. Once Sections Have Been Incubated with Peroxidase, Wash Sections with Wash Buffer. 2. Wipe Slides to Remove Excess Buffer. Add Enough Drops of AEC Chromogen/Substrate to Cover Tissue Sections. 3. Incubate for 5- 15 Minutes (at Room Temperature. 4. For Best Results, Observe Reaction Under a Microscope for Signal Development. 5. Once Desired Signal to Noise Ratio is Achieved, Stop Reaction by Washing Slides in DI H2O.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.