missing translation for 'onlineSavingsMsg'
Learn More

AF-353, MedChemExpress

Product Code. 16486078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486078 10 mg 10mg
16472217 50 mg 50mg
16428687 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486078 Supplier Medchem Express Supplier No. HY14483
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AF-353 (Ro-4) is a potent, selective and orally bioavailable P2X3/P2X2/3 receptor antagonist, with a pIC50 of 8.0 for both human and rat P2X3, and with a pIC50 of 7.3 for human P2X2/3.

AF-353 (Ro-4) is a potent, selective and orally bioavailable P2X3/P2X2/3 receptor antagonist, with a pIC50 of 8.0 for both human and rat P2X3, and with a pIC50 of 7.3 for human P2X2/3.

Specifications

Chemical Name or Material AF-353
CAS 865305-30-2
Color Brown
Purity Grade Notes Research
Molecular Formula C14H17IN4O2
Quantity 10 mg
Synonym Ro-4
Solubility Information DMSO : 100 mg/mL (249.87 mM; Need ultrasonic)
SMILES CC(C)C1=C(OC2=CN=C(N)N=C2N)C=C(I)C(OC)=C1
Molecular Weight (g/mol) 400.21
Formula Weight 400.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.