missing translation for 'onlineSavingsMsg'
Learn More

AF-353, MedChemExpress

Product Code. 16428687
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16428687

Brand: Medchem Express HY14483

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AF-353 (Ro-4) is a potent, selective and orally bioavailable P2X3/P2X2/3 receptor antagonist, with a pIC50 of 8.0 for both human and rat P2X3, and with a pIC50 of 7.3 for human P2X2/3.

AF-353 (Ro-4) is a potent, selective and orally bioavailable P2X3/P2X2/3 receptor antagonist, with a pIC50 of 8.0 for both human and rat P2X3, and with a pIC50 of 7.3 for human P2X2/3.

Specifications

Chemical Name or Material AF-353
CAS 865305-30-2
Color Brown
Purity Grade Notes Research
Molecular Formula C14H17IN4O2
Quantity 5 mg
Synonym Ro-4
Solubility Information DMSO : 100 mg/mL (249.87 mM; Need ultrasonic)
SMILES CC(C)C1=C(OC2=CN=C(N)N=C2N)C=C(I)C(OC)=C1
Molecular Weight (g/mol) 400.21
Formula Weight 400.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.