missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B1, B2, G1, G2 and Ochratoxin A Kit 2 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30315872
Change view
Click to view available options
Quantity:
2 x 1 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315872 2 x 1 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315872 Supplier Dr. Ehrenstorfer Supplier No. DREK50000755AL
 Restricted Item
Estimated Shipment: 07-10-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Aflatoxin B1∣Aflatoxin B2∣Aflatoxin G1∣Aflatoxin G2∣Ochratoxin A
CAS 1162-65-8, 7220-81-7, 1165-39-5, 7241-98-7, 303-47-9
Analyte or Component Names Aflatoxin B1∣Aflatoxin B2∣Aflatoxin G1∣Aflatoxin G2∣Ochratoxin A
Shipping Condition -18°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 2 x 1 mL
Concentration or Composition (by Analyte or Components) Aflatoxin B1 2 μg/mL∣Aflatoxin B2 2 μg/mL∣Aflatoxin G1 2 μg/mL∣Aflatoxin G2 2 μg/mL∣Ochratoxin A 2 μg/mL
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.