missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B1, B2, G1, G2 and Ochratoxin A Kit 2 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318180
Change view
Click to view available options
Quantity:
2 x 5 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318180 2 x 5 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318180 Supplier Dr. Ehrenstorfer Supplier No. DREK50000756AL
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aflatoxin B1∣Aflatoxin B2∣Aflatoxin G1∣Aflatoxin G2∣Ochratoxin A
CAS 1162-65-8, 7220-81-7, 1165-39-5, 7241-98-7, 303-47-9
Analyte or Component Names Aflatoxin B1∣Aflatoxin B2∣Aflatoxin G1∣Aflatoxin G2∣Ochratoxin A
Shipping Condition -18°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 2 x 5 x 1 mL
Concentration or Composition (by Analyte or Components) Aflatoxin B1 2 μg/mL∣Aflatoxin B2 2 μg/mL∣Aflatoxin G1 2 μg/mL∣Aflatoxin G2 2 μg/mL∣Ochratoxin A 2 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.