missing translation for 'onlineSavingsMsg'
Learn More

AG 555, MedChemExpress

Product Code. 16476736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476736 Supplier Medchem Express Supplier No. HY15336
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AG 555 (Tyrphostin AG 555), a potent antiretroviral drug, is a potent and selective inhibitor of EGFR and blocks Cdk2 activation.

AG 555 (Tyrphostin AG 555), a potent antiretroviral drug, is a potent and selective inhibitor of EGFR and blocks Cdk2 activation.

Specifications

Chemical Name or Material AG 555
CAS 133550-34-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H18N2O3
Quantity 100 mg
Synonym Tyrphostin AG 555
Solubility Information DMSO : ≥ 100 mg/mL (310.21 mM)
SMILES O=C(NCCCC1=CC=CC=C1)/C(C#N)=C/C2=CC=C(O)C(O)=C2
Molecular Weight (g/mol) 322.36
Formula Weight 322.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.