missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Agate Comfort Grinding Jars

Use these grinding jars with Retsch™ ball mills. Retsch™ Agate Comfort Grinding Jars feature an O-type sealing for safe operation.

Brand:  RETSCH 01.462.0197

Code : 50

Additional Details : Weight : 1.20000kg

 View more versions of this product

Product Code. 10044901

  • 2330.00 € / Each

Estimated Shipment: 29-06-2023
to see stock.
Add to basket

Description

Description

  • Available in a variety of sizes from 12 to 500mL
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications

Specifications

Comfort Grinding Jar
80 mL
For PM 100, PM 200, PM 400 and S 100
Agate
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers