Learn More
AGER Antibody (CL10587) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07982AF405
Product Code. 30304453
  • 737.00 € / 0.10mL
Description
AGER Monoclonal antibody specifically detects AGER in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AGER
Monoclonal
Alexa Fluor 405
advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products
This antibody was developed using a synthetic peptide derived from Q15109, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
177
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10587
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Rat
Purified