missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGER Antibody (CL10591), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21177-100ul
This item is not returnable.
View return policy
Description
AGER Monoclonal antibody specifically detects AGER in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AGER | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10591 | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
| This antibody was developed against a Synthetic Peptide of Q15109. Actual immunogen sequence is proprietary. | |
| 100 μg | |
| Cancer | |
| 177 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction