missing translation for 'onlineSavingsMsg'
Learn More

Agmatine sulfate, MedChemExpress

Product Code. 16490739
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490739 1 g 1g
16471526 500 mg 500mg
16454556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490739 Supplier Medchem Express Supplier No. HY101238
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Agmatine sulfate exerts modulatory action at multiple molecular targets, such as neurotransmitter systems, ion channels and nitric oxide synthesis. It is an endogenous agonist at imidazoline receptor and a NO synthase inhibitor.

Agmatine sulfate exerts modulatory action at multiple molecular targets, such as neurotransmitter systems, ion channels and nitric oxide synthesis. It is an endogenous agonist at imidazoline receptor and a NO synthase inhibitor.

Specifications

Chemical Name or Material Agmatine sulfate
Physical Form Solid
CAS 2482-00-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C5H16N4O4S
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : ≥ 100 mg/mL (438.08 mM) ∣DMSO : 1 mg/mL (4.38 mM; ultrasonic and warming and heat to 80°C)
SMILES NC(NCCCCN)=N.O=S(O)(O)=O
Molecular Weight (g/mol) 228.27
Formula Weight 228.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.