missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ago2/eIF2C2 Rabbit anti-Mouse, Clone: 36, Novus Biologicals™
Rabbit Monoclonal Antibody
392.00 € - 578.00 €
Specifications
| Antigen | Ago2/eIF2C2 |
|---|---|
| Clone | 36 |
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry, Immunoprecipitation 1-4 ul/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18228517
|
Novus Biologicals
NBP2-90675-50UL |
50 μg |
392.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18287236
|
Novus Biologicals
NBP2-90675-100UL |
100 μg |
578.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ago2/eIF2C2 Monoclonal specifically detects Ago2/eIF2C2 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Ago2/eIF2C2 | |
| ELISA 1:5000-1:10000, Immunohistochemistry, Immunoprecipitation 1-4 ul/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10 | |
| Recombinant Mouse Ago2/eIF2C2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 36 | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| 0.2 um filtered solution in PBS | |
| 27161 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title