missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGPS Recombinant Rabbit Monoclonal Antibody (24GB6810)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | AGPS |
|---|---|
| Clone | 24GB6810 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276062
![]() |
Invitrogen™
MA556957 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275806
|
Invitrogen™
MA556958 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGPS Recombinant Monoclonal Antibody for Western Blot
This gene is a member of the FAD-binding oxidoreductase/transferase type 4 family. It encodes a protein that catalyzes the second step of ether lipid biosynthesis in which acyl-dihydroxyacetonephosphate (DHAP) is converted to alkyl-DHAP by the addition of a long chain alcohol and the removal of a long-chain acid anion. The protein is localized to the inner aspect of the peroxisomal membrane and requires FAD as a cofactor. Mutations in this gene have been associated with rhizomelic chondrodysplasia punctata, type 3 and Zellweger syndrome.Specifications
| AGPS | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| AGPS | |
| 5832437L22; 9930035G10Rik; AAG5; Adaps; Adap-s; ADAS; Adhaps; ADPS; Ads; Aging-associated gene 5 protein; aging-associated protein 5; AGPS; ALDHPSY; alkyl-DHAP synthase; alkyldihydroxyacetone phosphate synthase; alkyl-dihydroxyacetonephosphate synthase; alkyldihydroxyacetonephosphate synthase, peroxisomal; alkylglycerone phosphate synthase; alkylglycerone phosphate synthase-like; alkylglycerone-phosphate synthase; AW123847; bs2; RCDP3 | |
| AGPS | |
| Primary | |
| Affinity chromatography |
| 24GB6810 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00116, Q9EQR2 | |
| 84114, 8540 | |
| Recombinant protein of human Alkyl-DHAP synthase (aa 71-182). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title