missing translation for 'onlineSavingsMsg'
Learn More

AIF1 / Iba1 (Microglia Marker) Mouse, Clone: AIF1/1909, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 478.00 €

Specifications

Antigen AIF1 / Iba1 (Microglia Marker)
Clone AIF1/1909
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15949937
missing translation for 'viewDocuments'
EnquireBio
199-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
15959937
missing translation for 'viewDocuments'
EnquireBio
199-MSM1-P1
100 μg
ONLINE EXCLUSIVE PRICE
554.40 €
478.00 € / 100µg
Save 76.40 €
13% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

AIF1 / Iba1 (Microglia Marker)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Purified fragment of human recombinant AIF1 protein (apprx. aa 1-146)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
17kDa
AIF1/1909
Immunohistochemistry
Unconjugated
RUO
199
IgG1 κ
Protein A/G purified from Bioreactor concentrate.
Cytoplasmic and Cell Surface
Lymph Node, Tonsil or Kidney
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.