missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99233-100ul
This item is not returnable.
View return policy
Description
AIF1L Polyclonal antibody specifically detects AIF1L in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| AIF1L | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| allograft inflammatory factor 1-like, C9orf58, chromosome 9 open reading frame 58, IBA2FLJ12783, ionized calcium binding adapter molecule 2, Ionized calcium-binding adapter molecule 2, MGC29466 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human AIF1L. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 83543 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction