missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 487.00 €
Specifications
| Antigen | AK7 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657800
|
Novus Biologicals
NBP2-92176-0.02ml |
0.02 mL |
216.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633160
|
Novus Biologicals
NBP2-92176-0.1ml |
0.1 mL |
487.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AK7 Polyclonal antibody specifically detects AK7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| AK7 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 122481 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| adenylate kinase 7, EC 2.7.4.3, FLJ32864, putative adenylate kinase 7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human AK7 (NP_689540.2). MAEEEETAALTEKVIRTQRVFINLLDSYSSGNIGKFLSNCVVGASLEEITEEEEEEDENKSAMLEASSTKVKEGTFQIVGTLSKPDSPRPDFAVETYSAISREDLLMRLLECDVIIYNITESSQQMEEAIWAVSALSEEVSHFEKRKLFILLSTVMTWARSKALDPEDSEVPFTEEDYRRRKSHPNFLDHINAEKMVLKF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title