missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38713-25ul
This item is not returnable.
View return policy
Description
AKAP5 Polyclonal specifically detects AKAP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AKAP5 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P24588 | |
| AKAP5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LDIQTQTPLNDQATKAKSTQDLSEGISRKDGDEVCESNVSNSITSGEKVISVELGLDNGHSAIQTGTLILEEIE | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| A kinase (PRKA) anchor protein 5, AKAP 79, AKAP7979kDa, A-kinase anchor protein 79 kDa, A-kinase anchoring protein 75/79, cAMP-dependent protein kinase regulatory subunit II high affinity bindingprotein, cAMP-dependent protein kinase regulatory subunit II high affinity-bindingprotein, H21 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9495 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction