missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKR1C4 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149274
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the bioreduction of chlordecone, a toxic organochlorine pesticide, to chlordecone alcohol in liver. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14.
Specifications
| AKR1C4 | |
| Polyclonal | |
| Unconjugated | |
| AKR1C4 | |
| 3-alpha-HSD; 3-alpha-HSD1; 3-alpha-hydroxysteroid dehydrogenase type I; AKR1C4; Aldo-keto reductase family 1 member C4; aldo-keto reductase family 1, member C4; C11; CDR; CHDR; Chlordecone reductase; chlordecone reductase; 3-alpha hydroxysteroid dehydrogenase, type I; dihydrodiol dehydrogenase 4; DD4; DD-4; Dihydrodiol dehydrogenase 4; dihydrodiol dehydrogenase isozyme DD4; HAKRA; HGNC:387; MGC22581; type I 3-alpha-hydroxysteroid dehydrogenase | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 1109 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P17516 | |
| AKR1C4 | |
| E. coli-derived recombinant human Aldo-keto Reductase 1C4/AKR1C4, Met1-Tyr323. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction