missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT Antibody (0002) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
186.00 € - 444.00 €
Specifications
| Antigen | AKT |
|---|---|
| Clone | 0002 |
| Dilution | Western Blot 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646494
|
Novus Biologicals
NBP3-23519-100ul |
100 μL |
444.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644314
|
Novus Biologicals
NBP3-23519-20ul |
20 μL |
186.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKT Monoclonal antibody specifically detects AKT in Human samples. It is validated for Western BlotSpecifications
| AKT | |
| Western Blot 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the residues around Ser308 of Human phospho-AKT. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0002 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cellular Signaling, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 207 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title