missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
241.00 € - 530.00 €
Specifications
| Antigen | AKT2 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630569
|
Novus Biologicals
NBP3-05341-100ul |
100 μg |
530.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694669
|
Novus Biologicals
NBP3-05341-20ul |
20 μg |
241.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKT2 Polyclonal antibody specifically detects AKT2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| AKT2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Protein Kinase, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 208 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 381-481 of human AKT2 (NP_001617.1). LAGLLKKDPKQRLGGGPSDAKEVMEHRFFLSINWQDVVQKKLLPPFKPQVTSEVDTRYFDDEFTAQSITITPPDRYDSLGLLELDQRTHFPQFSYSASIRE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title