missing translation for 'onlineSavingsMsg'
Learn More

Alagebrium chloride, MedChemExpress

Product Code. 16480996
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480996 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16480996 Supplier Medchem Express Supplier No. HY106024B
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alagebrium chloride (ALT711) is an advanced glycation end product (AGE) inhibitor.

Alagebrium chloride (ALT711) is an advanced glycation end product (AGE) inhibitor.

Specifications

Chemical Name or Material Alagebrium chloride
Physical Form Solid
CAS 341028-37-3
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C13H14ClNOS
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym ALT711
Solubility Information H2O : 50 mg/mL (186.73 mM; Need ultrasonic) ∣DMSO : ≥ 25 mg/mL (93.36 mM)
SMILES O=C(C1=CC=CC=C1)C[N+]2=CSC(C)=C2C.[Cl-]
Molecular Weight (g/mol) 267.77
Formula Weight 267.77
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.